missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red sodium salt, 0.04% w/v aq. soln.

Catalog No. AA42752AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42752AE 100 mL
AA42752AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42752AE Supplier Thermo Scientific Chemicals Supplier No. 042752AE

CAS: 62625-29-0 | C21H17O5S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62625-29-0
Molecular Formula C21H17O5S
MDL Number MFCD00001618

Specifications

Concentration 0.04% w/v aq. soln.
Formula Weight 404.42
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Cresol Red sodium salt
EINECSNumber 263-654-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only