missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red, pure, Indicator grade

CAS: 1733-12-6 | C21H18O5S | 382.43 g/mol

$51.72 - $522.67

Chemical Identifiers

CAS 1733-12-6
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005878
InChI Key OBRMNDMBJQTZHV-UHFFFAOYSA-N
PubChem CID 73013
ChEBI CHEBI:86218
IUPAC Name 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
SMILES CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151380050
SDS View Documents
Thermo Scientific Chemicals
151380050
5 g Glass bottle
Each for $51.72
Only null left
 
AC151380250
SDS View Documents
Thermo Scientific Chemicals
151380250
25 g Glass bottle
Each for $140.88
Only null left
 
AC151381000
SDS View Documents
Thermo Scientific Chemicals
151381000
100 g Glass bottle
Each for $522.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

1733-12-6
382.43
OBRMNDMBJQTZHV-UHFFFAOYSA-N
CHEBI:86218
CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
C21H18O5S
MFCD00005878
73013
4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol

Specifications

290.0°C
C21H18O5S
19, 91
Solubility in water: soluble. Other solubilities: soluble in dil. acids, dil. alkalies and ethanol,, insoluble in diethyl ether, benzene
Authentic
4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
73013
382.42
Glass bottle
from pH 7.0 (orange) to pH 8.8 (purple)
5 g
Cresol Red
1733-12-6
MFCD00005878
15, 2568
OBRMNDMBJQTZHV-UHFFFAOYSA-N
CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
382.43
CHEBI:86218
Pure
1500 min. (572nm, in 0.01N NaOH)
Brown/Red or Green
Powder
Videos
SDS
Documents

Documents

RUO – Research Use Only