missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresol Red, pure, Indicator grade

CAS: 1733-12-6 | C21H18O5S | 382.43 g/mol

Supplier:  Thermo Scientific Chemicals 151380250

 View more versions of this product

Catalog No. AC151380250

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Cresol Red
1733-12-6
MFCD00005878
15, 2568
OBRMNDMBJQTZHV-UHFFFAOYSA-N
CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
382.43
CHEBI:86218
Pure
1500 min. (572nm, in 0.01N NaOH)
Brown/Red or Green
Powder
290.0°C
C21H18O5S
19, 91
Solubility in water: soluble. Other solubilities: soluble in dil. acids, dil. alkalies and ethanol,, insoluble in diethyl ether, benzene
Authentic
4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
73013
382.42
Glass bottle
from pH 7.0 (orange) to pH 8.8 (purple)
25 g

Chemical Identifiers

1733-12-6
382.43
OBRMNDMBJQTZHV-UHFFFAOYSA-N
CHEBI:86218
CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1
C21H18O5S
MFCD00005878
73013
4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only