missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cresol Red Indicator, ≥83%, Honeywell Fluka™
Indicator
$380.26 - $380.26
Chemical Identifiers
| CAS | 1733-12-6 |
|---|---|
| Molecular Formula | C21H18O5S |
| Molecular Weight (g/mol) | 382.43 |
| MDL Number | MFCD00005878 |
| InChI Key | OBRMNDMBJQTZHV-UHFFFAOYSA-N |
| Synonym | o-Cresolsulfonphthalein |
| PubChem CID | 73013 |
| ChEBI | CHEBI:86218 |
| IUPAC Name | 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol |
| SMILES | CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1 |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1733-12-6 | |
| 382.43 | |
| OBRMNDMBJQTZHV-UHFFFAOYSA-N | |
| 73013 | |
| 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol |
| C21H18O5S | |
| MFCD00005878 | |
| o-Cresolsulfonphthalein | |
| CHEBI:86218 | |
| CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1 |
Specifications
| 1733-12-6 | |
| MFCD00005878 | |
| 343399 | |
| OBRMNDMBJQTZHV-UHFFFAOYSA-N | |
| 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol | |
| 73013 | |
| 0 to 5% | |
| Glass Bottle | |
| Brown/Red | |
| 25 g | |
| Cresol Red |
| C21H18O5S | |
| NONH for all modes of transport | |
| o-Cresolsulfonphthalein | |
| CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1 | |
| 382.43 | |
| CHEBI:86218 | |
| 382.43g/mol | |
| 0.998g/cm3 | |
| 7.2 to 8.8 (yellow to red) | |
| Powder |
Safety and Handling
H315 + H319 + H335
P261 + P264 + P271 + P280 + P302 + P352+ P304 + P340 + P305 + P351 + P338
Warning
EINECSNumber : 217-064-2