missing translation for 'onlineSavingsMsg'
Learn More

Cresol Red Indicator, ≥83%, Solstice

Catalog No. 60046142
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046142 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60046142 Supplier Solstice Supplier No. 3265325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1733-12-6
Molecular Formula C21H18O5S
Molecular Weight (g/mol) 382.43
MDL Number MFCD00005878
InChI Key OBRMNDMBJQTZHV-UHFFFAOYSA-N
Synonym o-Cresolsulfonphthalein
PubChem CID 73013
ChEBI CHEBI:86218
IUPAC Name 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-methylphenol
SMILES CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

Loss on Drying 0 to 5%
Packaging Glass Bottle
UN Number NONH for all modes of transport
Beilstein 343399
Density 0.998g/cm3
Color Brown/Red
pH 7.2 to 8.8 (yellow to red)
Quantity 25 g
Physical Form Powder
Formula Weight 382.43g/mol
Chemical Name or Material Cresol Red
Show More Show Less
Hazard Category Warning
Hazard Statement H315 + H319 + H335
Precautionary Statement P261 + P264 + P271 + P280 + P302 + P352+ P304 + P340 + P305 + P351 + P338
EINECSNumber 217-064-2