missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CREB5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149961
Description
CREB5 Polyclonal Antibody for Western Blot, ICC/IF, IHC
The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
| CREB5 | |
| Polyclonal | |
| Unconjugated | |
| Creb5 | |
| cAMP response element-binding protein CRE-BPa; cAMP responsive element binding protein 5; cAMP-response element-binding protein A; cAMP-responsive element-binding protein 5; CREB5; CREB-5; CREBPA; CRE-BPa; Cyclic AMP-responsive element-binding protein 5; D430026C09Rik; RGD1566107 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 231991, 500131, 9586 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q02930, Q8K1L0 | |
| Creb5 | |
| Recombinant Cyclic AMP Response Element Binding Protein 5 (CREB5) (Gln308-Leu508). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |