missing translation for 'onlineSavingsMsg'
Learn More

Creatinol Phosphate 98.0+%, TCI America™

Supplier:  TCI America C27675G

 View more versions of this product

Catalog No. C27675G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Creatinol Phosphate
244°C
244°C
MFCD00868297
2-(1-Methylguanidino)ethyl Dihydrogen Phosphate
CN(CCOP(O)(O)=O)C(N)=N
197.13
197.13
Crystalline Powder
6903-79-3
White
C4H12N3O4P
5 g
FOIPWTMKYXWFGC-UHFFFAOYSA-N
[2-(N-methylcarbamimidamido)ethoxy]phosphonic acid
23342
≥98.0% (HPLC,T)

Chemical Identifiers

6903-79-3
197.13
FOIPWTMKYXWFGC-UHFFFAOYSA-N
23342
CN(CCOP(O)(O)=O)C(N)=N
C4H12N3O4P
MFCD00868297
2-(1-Methylguanidino)ethyl Dihydrogen Phosphate
[2-(N-methylcarbamimidamido)ethoxy]phosphonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MF3600500

TSCA : No

SDS
Documents

Documents

Promotions

Promotions