CAS RN 6903-79-3
CAS RN 6903-79-3
IUPAC Name:
[2-(N-methylcarbamimidamido)ethoxy]phosphonic acid
Synonyms:
aplodan creatinol O-phosphate creatinol phosphate, magnesium salt (2:1) magnesium salt (2:1) creatinol phosphate creatinol-o-phosphate creatinolfosfat creatinolo O-fosfato creatinol-O-phosphate creatinol phosphate creatinolfosfato
Molecular Weight (g/mol):
197.13
Molecular Formula:
C4H12N3O4P
InChi Key:
FOIPWTMKYXWFGC-UHFFFAOYSA-N
SMILES:
CN(CCOP(O)(O)=O)C(N)=N
1
–
1
of
1
results
1
Creatinol Phosphate 98.0+%, TCI America™
CAS: 6903-79-3 Molecular Formula: C4H12N3O4P Molecular Weight (g/mol): 197.13 MDL Number: MFCD00868297 InChI Key: FOIPWTMKYXWFGC-UHFFFAOYSA-N Synonym: 2-(1-Methylguanidino)ethyl Dihydrogen Phosphate PubChem CID: 23342 IUPAC Name: [2-(N-methylcarbamimidamido)ethoxy]phosphonic acid SMILES: CN(CCOP(O)(O)=O)C(N)=N