missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatinol Phosphate 98.0+%, TCI America™
Supplier: TCI America C276725G
Specifications
| Creatinol Phosphate | |
| 244°C | |
| 244°C | |
| MFCD00868297 | |
| 2-(1-Methylguanidino)ethyl Dihydrogen Phosphate | |
| CN(CCOP(O)(O)=O)C(N)=N | |
| 197.13 | |
| 197.13 | |
| Crystalline Powder |
| 6903-79-3 | |
| White | |
| C4H12N3O4P | |
| 25 g | |
| FOIPWTMKYXWFGC-UHFFFAOYSA-N | |
| [2-(N-methylcarbamimidamido)ethoxy]phosphonic acid | |
| 23342 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 6903-79-3 | |
| 197.13 | |
| FOIPWTMKYXWFGC-UHFFFAOYSA-N | |
| 23342 | |
| CN(CCOP(O)(O)=O)C(N)=N |
| C4H12N3O4P | |
| MFCD00868297 | |
| 2-(1-Methylguanidino)ethyl Dihydrogen Phosphate | |
| [2-(N-methylcarbamimidamido)ethoxy]phosphonic acid |
Safety and Handling
RTECSNumber : MF3600500
TSCA : No