missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatinine, NF, 98.5-102%, Spectrum™ Chemical
C1403, 60-27-5, C4H7N3O
Supplier: Spectrum Chemical Mfg Cor C140312KGBL
Description
Spectrum™ Chemical Creatinine, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 60-27-5 | |
| C4H7N3O | |
| DDRJAANPRJIHGJ-UHFFFAOYSA-N | |
| 2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one | |
| 98.5 to 102% | |
| 0.2% | |
| Poly Pail |
| 100% | |
| 12 kg | |
| CN1CC(=O)N=C1N | |
| 113.12 | |
| NF | |
| 3% |
Chemical Identifiers
| 60-27-5 | |
| 113.12 | |
| 2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one |
| C4H7N3O | |
| DDRJAANPRJIHGJ-UHFFFAOYSA-N | |
| CN1CC(=O)N=C1N |