missing translation for 'onlineSavingsMsg'
Learn More

Medisca Creatine (Monohydrate) (Dietary Supplement Grade)

Catalog No. 14186057
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186057 500 g
14185481 100 g
14185511 250 g
14186220 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14186057 Supplier Medisca Supplier No. 083508

Creatine (Monohydrate) (Dietary Supplement Grade) is a white, odorless, crystalline powder that is sparingly soluble in water.

Creatine (Monohydrate) is a white, odorless, crystalline powder that is sparingly soluble in water.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): Creatine hydrate, Creatine monohydrate

Specifications

Chemical Name or Material Creatine (Monohydrate) (Dietary Supplement Grade)
Packaging Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 6020-87-7
Type Drug Standard
Molecular Weight (g/mol) 149.1489
Quantity 500 g
Percent Purity 99 to 102 %
Grade Ungraded