missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF68 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
CPSF6 is one subunit of a cleavage factor required for 3′ RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3′ end processing complex and facilitates the recruitement of other processing factors. The cleavage factor complex is composed of four polypeptides.The protein encoded by this gene is one subunit of a cleavage factor required for 3′ RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3′ end processing complex and facilitates the recruitement of other processing factors. The cleavage factor complex is composed of four polypeptides. This gene encodes the 68kD subunit. It has a domain organization reminiscent of spliceosomal proteins.
Specifications
Specifications
| Antigen | CPSF68 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | CPSF6 |
| Gene Accession No. | Q16630, Q6NVF9 |
| Gene Alias | Cleavage and polyadenylation specific factor 6, CFIM, CFIM68, HPBRII-4, HPBRII-7 |
| Gene Symbols | CPSF6 |
| Show More |