missing translation for 'onlineSavingsMsg'
Learn More

CPM, Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89019989
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89019989 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89019989 Supplier Abnova Corporation Supplier No. H00001368B01P
Only null left

Mouse Polyclonal Antibody

Sequence: MDFPCLWLGLLLPLVAALDFNYHRQEGMEAFLKTVAQNYSSVTHLHSIGKSVKGRNLWVLVVGRFPKEHRIGIPEFKYVANMHGDETVGRELLLHLIDYLVTSDGKDPEITNLINSTRIHIMPSMNPDGFEAVKKPDCYYSIGRENYNQYDLNRNFPDAFEYNNVSRQPETVAVMKWLKTETFVLSANLHGGALVASYPFDNGVQATGALYSRSLTPDDDVFQYLAHTYASRNPNMKKGDECKNKMNFPNGVTNGYSWYPLQGGMQDYNYIWAQCFEITLELSCCKYPREEKLPSFWNNNKASLIEYIKQVHLGVKGQVFDQNGNPLPNVIVEVQDRKHICPYRTNKYGEYYLLLLPGSYIINVTVPGHDPHITKVIIPEKSQNFSALKKDILLPFQGQLDSIPVSNPSCPMIPLYRNLPDHSAATKPSLFLFLVSLLHIFFK

Specifications

Antigen CPM
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene carboxypeptidase M
Gene Accession No. NM_001005502.1
Gene Alias -
Gene Symbols CPM
Host Species Mouse
Immunogen CPM (NP_001005502.1, 1 a.a. ∼ 443 a.a) full-length human protein.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1368
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Show More Show Less