missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CPM (7-Diethylamino-3-(4'-Maleimidylphenyl)-4-Methylcoumarin)

Catalog No. d346
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D346 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D346 Supplier Invitrogen™ Supplier No. D346

The thiol-reactive coumarin, CPM is very weakly fluorescent until reacted with thiols producing a conjugate with excitation/emission maxima of ∼384/470 nm.

The thiol-reactive coumarin, CPM is very weakly fluorescent until reacted with thiols producing a conjugate with excitation/emission maxima of ∼384/470 nm.

Specifications

Quantity 25 mg
Product Type CPM
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye Methylcoumarin
Label Type Classic Dyes
Emission 470 nm
Excitation 384 nm

For Research Use Only. Not for use in diagnostic procedures.