missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CPLX1/2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA5149404
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5149404 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5149404 Supplier Invitrogen™ Supplier No. PA5149404

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Complexins (1 through 4) are small cytosolic proteins that bind tightly to a complex of SNARE proteins to regulate synaptic vesicle membrane fusion via its interaction withsynaptotagmin. Complexins activate the quick release of neurotransmitters, triggered by calcium release. They also regulate the spontaneous release of neurotransmitters. Complexins are evolutionarily conserved in mammals, Drosophila, and Caenorhabditis elegans. They are divided into four structural domains: flexible N- and C-terminal domains, an accessory domain, and a central alpha-helical domain. The N-terminal domain plays a role in activation of fast synchronous release in murine neurons and in isolated chromaffin cells. The C-terminal domain binds to phospholipids and is important for regulating spontaneous release in neuronal cultures and suppressing Ca2+-independent fusion in vitro. The accessory domain is necessary for regulating spontaneous release and the central domain is required for all functions.

Specifications

Antigen CPLX1/2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene Cplx1
Gene Accession No. O14810, P63040, P63041, P84086, P84087, Q6PUV4
Gene Alias 921-L; 921-S; AI413745; AW492120; complexin 1; complexin 2; complexin I; Complexin II; complexin-1; Complexin-2; Cplx1; Cplx2; CPX I; CPX II; CPX1; CPX2; CPX-2; CPX-I; Hfb1; synaphin 1; synaphin 2; synaphin-1; Synaphin-2
Gene Symbols Cplx1, Cplx2
Host Species Goat
Immunogen E. coli-derived recombinant human Complexin-1, Met1-Lys134.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10814, 10815, 116657, 12889, 12890, 64832
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less