missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon, +12 Mesh to 1 in., Lump, 96%75, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Irregular Lumps/Pieces |
| Quantity | 2.5 kg |
| Percent Purity | 96.75% |
| Grade | Not Available |
| Packaging | Poly Bottle |
| Chemical Name or Material | Silicon, +12 Mesh to 1 in., Lump |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.