Learn More
Applied Biosystems™ CPG (Controlled Pore Glass) Synthesis Column, 1.0 μmol, 1000 Angstrom Pore Size, T
Catalog No. 401441
Encompass
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Column
Catalog No. Quantity
401441 1 Column
1 missing translation for 'options'
Catalog No. 401441 missing translation for 'mfr' Applied Biosystems™ missing translation for 'supplierNo' LS401441
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The 1.0μmol T Controlled Pore Glass (CPG) columns are suitable for the synthesis of any length oligonucleotide. These columns deliver high-quality oligonucleotides.
These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.
  • Proprietary design and manufacturing methods ensure consistency and high quality
  • Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
  • The 1,000 angstrom pore size allows synthesis of oligonucleotides of varying length
Specifications
Packaging Type Shipped at Room Temperature
Quantity 1 Column
For Research Use Only. Not for use in diagnostic procedures.