missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guardian™ CP-5, Chemical Resistant Smooth Finish 5 mil Butyl Gloves
USA made, 14 in. butyl gloves offer protection against a variety of hazardous chemicals.
$69.10 - $86.36
Specifications
| Color | Black |
|---|---|
| Hand | Specific |
| Latex Free | Yes |
| Material | Butyl |
| Powdered | Powdered |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19-321-184
|
Guardian™
GGL50501 |
X-Small |
Pair for $69.10
|
|
|||||
|
19-321-185
|
Guardian™
GGL50502 |
Small |
Pair for $69.10
|
|
|||||
|
19-321-186
|
Guardian™
GGL50503 |
Medium |
Pair for $69.10
|
|
|||||
|
19-321-187
|
Guardian™
GGL50504 |
Large |
Pair for $69.10
|
|
|||||
|
19-321-188
|
Guardian™
50505 |
X-Large |
Pair for $86.36
|
|
|||||
Description
- Butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with smooth texture
- Protects against wide range of hazardous chemicals
- 5 mil thickness, 14 in. length
- Made in the USA
Specifications
| Black | |
| Yes | |
| Powdered | |
| 0.12 mm | |
| Smooth | |
| 14 in. |
| Specific | |
| Butyl | |
| 5 mil | |
| Gauntlet | |
| Unlined | |
| 35.5 cm |