missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guardian™ CP-5, Chemical Resistant Smooth Finish 5 mil Butyl Gloves
USA made, 14 in. butyl gloves offer protection against a variety of hazardous chemicals.
Supplier: Guardian™ GGL50501
Description
- Butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with smooth texture
- Protects against wide range of hazardous chemicals
- 5 mil thickness, 14 in. length
- Made in the USA
Specifications
| Black | |
| Yes | |
| Powdered | |
| 5 mil | |
| Gauntlet | |
| Unlined | |
| 14 in. |
| Specific | |
| Butyl | |
| X-Small | |
| 0.12 mm | |
| Smooth | |
| 50501 | |
| 35.5 cm |