missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COX6A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564017 Supplier Invitrogen™ Supplier No. PA564017
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: VYLKSHHGEH ERPEFIAYPH LRIRTK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 85%.

This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.

Specifications

Antigen COX6A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cox6a1
Gene Accession No. P12074
Gene Alias CMTRID; COX VIa-L; COX6A; Cox6a1; Cox6al; COXVIA2; Cytochrome c oxidase polypeptide VIa-liver; cytochrome c oxidase subunit 6A1; cytochrome c oxidase subunit 6A1, mitochondrial; Cytochrome c oxidase subunit SSG; cytochrome c oxidase subunit VIa; Cytochrome c oxidase subunit VIa (liver); cytochrome C oxidase subunit VIa homolog; cytochrome c oxidase subunit VIa polypeptide 1; cytochrome c oxidase subunit VIA-liver; cytochrome c oxidase, subunit VIa, polypeptide 1; cytochrome oxidase subunit VIa; cytochrome-c oxidase; ns:zf-e250; subunit VIaL (liver-type); Unknown (protein for MGC:140155); VIA L; VIaL; zgc:103782
Gene Symbols Cox6a1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COX6A1. Recombinant protein control fragment (Product #RP-104533).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1337
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less