missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Covidien Renewables™ Sharps Containers
Manufactured using recycled plastic
$44.42 - $1,013.42
Specifications
Certifications/Compliance | Meet or exceed the OSHA Bloodborne Pathogens Standard and conform to NIOSH/CDC reco mmendations. |
---|---|
Material | Recycled Plastic |
Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity | |||||
22-557-178
|
Covidien
8507MW |
5 qt. |
Case of 20 Each for $487.55
|
|
22-031349
|
Covidien
8900MW |
1 qt. |
Case of 100 Each for $1,013.42
|
|
22-037-965
|
Covidien
8537MW |
3 gal. |
Each for $44.42
Case of 10 Each for $444.20 |
|
Description
Meet or exceed the OSHA Bloodborne Pathogens Standard and conform to NIOSH/CDC recommendations.
Specifications
Meet or exceed the OSHA Bloodborne Pathogens Standard and conform to NIOSH/CDC reco mmendations. | |
Recycled Plastic |