Learn More
GSC Go Science Crazy Cover Plates
Catalog No. S24590
Encompass
Change view
Click to view available options
Dimensions (L x W):
4.5 x 4.5 in.
Catalog No. Dimensions (L x W)
S24590 4.5 x 4.5 in.
1 options
Catalog No. S24590 Supplier GSC Go Science Crazy Supplier No. S50346
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Used to cover beaker and flasks. GSC International Cover Plates are made of glass.
  • Features ground edges
  • Autoclavable
  • Keeps the content of flask free from contaminants
  • Available in different sizes
Specifications
Description Cover Plate
Material Glass
Shape Square
Dimensions (L x W) 4.5 x 4.5 in.