missing translation for 'onlineSavingsMsg'
Learn More

Coumarin-6-sulfonyl chloride, 97%

Catalog No. p-7036469
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5476703 1 g
AAH5476706 5 g
AAH5476714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5476703 Supplier Thermo Scientific Chemicals Supplier No. H5476703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10543-42-7 | C9H5ClO4S | 244.65 g/mol

Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 10543-42-7
Molecular Formula C9H5ClO4S
Molecular Weight (g/mol) 244.65
MDL Number MFCD01941320
InChI Key HQIPMBGUDSOVEA-UHFFFAOYSA-N
Synonym 2-oxo-2h-chromene-6-sulfonyl chloride, coumarin-6-sulfonyl chloride, 6-cs-cl, coumarin-6-sulphonyl chloride, 2-oxo-2h-chromene-6-sulphonyl chloride, 2-oxo-2h-1-benzopyran-6-sulfonyl chloride, 6-chlorosulfonyl chromen-2-one, pubchem5506, c6scl, timtec-bb sbb002796
PubChem CID 2735833
IUPAC Name 2-oxochromene-6-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C2OC(=O)C=CC2=C1

Specifications

Quantity 1 g
UN Number UN3261
Beilstein 196167
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 244.65
Percent Purity 97%
Chemical Name or Material Coumarin-6-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only