CAS RN 10543-42-7
IUPAC Name:
2-oxo-2H-chromene-6-sulfonyl chloride
Synonyms:
2-oxochromene-6-sulfonyl chloride
Molecular Weight (g/mol):
244.65
Molecular Formula:
C9H5ClO4S
InChi Key:
HQIPMBGUDSOVEA-UHFFFAOYSA-N
SMILES:
ClS(=O)(=O)C1=CC=C2OC(=O)C=CC2=C1
1
–
1
of
1
results
1
Promotions Available
Coumarin-6-sulfonyl chloride, 97%
Coumarin-6-sulfonyl chloride, 97%
CAS: 10543-42-7 Molecular Formula: C9H5ClO4S Molecular Weight (g/mol): 244.65 MDL Number: MFCD01941320 InChI Key: HQIPMBGUDSOVEA-UHFFFAOYSA-N Synonym: 2-oxo-2h-chromene-6-sulfonyl chloride,coumarin-6-sulfonyl chloride,6-cs-cl,coumarin-6-sulphonyl chloride,2-oxo-2h-chromene-6-sulphonyl chloride,2-oxo-2h-1-benzopyran-6-sulfonyl chloride,6-chlorosulfonyl chromen-2-one,pubchem5506,c6scl,timtec-bb sbb002796 PubChem CID: 2735833 IUPAC Name: 2-oxochromene-6-sulfonyl chloride SMILES: ClS(=O)(=O)C1=CC=C2OC(=O)C=CC2=C1