missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 153 98.0+%, TCI America™

Catalog No. C2858200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2858200MG 200 mg
C28581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2858200MG Supplier TCI America Supplier No. C2858200MG
Only null left

Chemical Identifiers

CAS 53518-18-6
Molecular Formula C16H14F3NO2
Molecular Weight (g/mol) 309.288
MDL Number MFCD00041843
InChI Key VSSSHNJONFTXHS-UHFFFAOYSA-N
Synonym coumarin 153, pilot 495, c 6f, coumarin 540a, 1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizin-11-one, 2,3,6,7-tetrahydro-9-trifluoromethyl, 2,3,6,7-tetrahydro-9-trifluoromethyl-1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizin-11-one, 9-trifluoromethyl-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinolin-11-one, 8-trifluoromethyl-2,3,5,6-tetrahydro-1h,4h-11-oxa-3a-aza-benzo de anthracen-10-one
PubChem CID 72652
ChEBI CHEBI:51773
SMILES C1CC2=C3C(=C4C(=C2)C(=CC(=O)O4)C(F)(F)F)CCCN3C1

Specifications

Melting Point 166°C
Color Yellow
Quantity 200 mg
Formula Weight 309.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 153