accessibility menu, dialog, popup

UserName

CAS RN 53518-18-6

IUPAC Name: 6-(trifluoromethyl)-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms: coumarin 153
Molecular Weight (g/mol): 309.29
Molecular Formula: C16H14F3NO2
InChi Key: VSSSHNJONFTXHS-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=O)OC2=C3CCCN4CCCC(C=C12)=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results