Learn More
Cottonseed Oil, NF, Spectrum™ Chemical
CO145, 8001-29-4
$341.97 - $9314.64
Chemical Identifiers
CAS 8001-29-4
View More Specs
Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601863
View Documents
Spectrum Chemical Mfg Cor
CO145500MLGL
500 mL Amber Glass Bottle
Each for $341.97
Only null left
18601864
View Documents
Spectrum Chemical Mfg Cor
CO1454LTGL
4 L Amber Glass Bottle
Each for $1,028.12
Only null left
18601865
View Documents
Spectrum Chemical Mfg Cor
CO14520LT
20 L Poly Drum
Each for $3,037.06
Only null left
18601866
View Documents
Spectrum Chemical Mfg Cor
CO145200LTBL
200 L Drum
Each for $9,314.64
Only null left
Description
Description
Spectrum™ Chemical Cottonseed Oil, NF is used as a solvent and vehicle for injections, as an emollient vehicle for other medications and orally as a mild cathartic. It can also be used as a tablet binder for stable oral pharmaceutical powders. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Chemical Identifiers
8001-29-4
Specifications
8001-29-4
Amber Glass Bottle
NF
100%
500 mL
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'