missing translation for 'onlineSavingsMsg'
Learn More

(-)-Cotinine Solution, Drug Standard, 1.0 mg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017156
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017156 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017156 Supplier Merck Emd Millipore Supplier No. C04301ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(−)-Cotinine is a major metabolite of nicotine, widely used as a biochemical marker for to evaluate the exposure to environmental tobacco smoke, for the validation of abstinence smoking and to determine the percentage of active smoking behavior.

Specifications

Packaging Ampule
Concentration 1.0 mg/mL in methanol
MDL Number MFCD00077696
Quantity 1 mL
Grade Drug Standard