missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Costar™ Transwell™ Inserts and Dishes
Applications include co-culture, drug transport, in vitro fertilization, and microbial pathogenesis studies.
$1907.21 - $1907.21
Specifications
| Bottom Shape | Round |
|---|---|
| Culture Area | 44 cm2 |
| Sterility | Sterile |
| Surface Treatment | Cell Culture |
| Type | Cell Culture Insert |
| Catalog Number | Mfr. No. | Supportive Membrane | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Supportive Membrane | Price | Quantity | |||||
|
07-200-173
|
Corning™
3420 |
3.0 μm |
Case of 12 Each for $1,907.21
|
|
|||||
Description
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
You can use cell culture inserts to:
- Produce a cell culture environment that closely resembles an in vivo state
- Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
- Pore sizes ranging from 0.4 μm to 8 μm diameters
- Treated for optimal cell attachment
- Supplied in multiple well plates
- Membrane must be stained for cell visibility
- Sterile
Order Info
Individually wrapped. Twelve inserts (with dishes) per case.
Specifications
| Round | |
| Sterile | |
| Cell Culture Insert | |
| 75 mm |
| 44 cm2 | |
| Cell Culture | |
| 9 mL |