missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Costar™ Netwell™ Inserts

Description
Netwell Inserts have polyester (PET) mesh bottoms attached to a polystyrene ring or housing. They are used as tissue carriers, supports and strainers for culture of small organs, tissue slices, or explants at the air-media interface. They can be used to coarse filter tissue homogenates, cell suspensions, or microcarriers. Accessories allow them to be used as a handy carrier for immunocytochemical staining of tissue culture slices.
Specifications
Specifications
| Array | 3 x 4 |
| Closure Included | Yes |
| Color | Clear |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Type | Cell Culture Insert |
| Well Count | 12 |
| For Use With (Application) | Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers |
| No. per Case | 48/Cs. |
| Show More |