missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ HTS Transwell™ Cell Culture Plate
GREENER_CHOICE

Catalog No. 07200399
Change view
Click to view available options
Well Count:
24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Count
07200399 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200399 Supplier Corning™ Supplier No. 4395
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tissue-culture treated

  • For use as reservoir tray with HTS Transwell Permeable Supports
  • TC-treated for cell attachment
  • Same footprint as standard 24 well plate but with single rectangular well

Specifications

Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Cell Culture
Type Cell Culture Plate
Well Count 24
Includes Lid
No. per Case 48/Cs.
No. per Pack 12/Pk.
Quantity 48/Pk.
Show More Show Less