missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ 24-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile
GREENER_CHOICE

Catalog No. 09761146
Change view
Click to view available options
Diameter (Metric):
1.6 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
09761146 1.6 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09761146 Supplier Corning™ Supplier No. C3526
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic

Specifications

Array 4 x 6
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 0.57 mL (Well)
Color Clear
Culture Area 1.9 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Cell Culture
Type Multi-well Cell Culture Plate
Volume (Metric) Working 0.5 mL
Well Count 24
Includes Lid
No. per Case 50/Cs.
No. per Pack 1/Pk.
Packaging Individually Wrapped
Quantity 50/Cs.
Diameter (Metric) 1.6 cm
Show More Show Less