missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Corticotropin-Releasing Factor, human, rat

Catalog No. AAJ66835EXD
Change view
Click to view available options
Quantity:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66835EXD 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66835EXD Supplier Thermo Scientific Chemicals Supplier No. J66835EXD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Molecular Weight (g/mol) 4757.44
Quantity 0.5mg
Packaging Plastic bottle
Form Solid

RUO – Research Use Only