Learn More
Invitrogen™ Corticosterone Competitive ELISA Kit
ELISA
$948.00 - $948.00
Specifications
| Assay Range | 78.125 to 10,000 pg/mL |
|---|---|
| Assay Sensitivity | 18.6 pg/mL |
| Description | Corticosterone Competitive ELISA Kit |
| Conjugate | HRP |
| Detection Method | Fluorescence |
Description
The Corticosterone ELISA quantitates coritcosterone in serum, plasma, urine, dried fecal extracts or cell culture medium. The assay will exclusively recognize both natural and recombinant coritcosterone. Principle of the method The corticosterone solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CORTICOSTERONE is an adrenocortical steroid that has modest but significant activities as a mineralocorticoid and a glucocorticoid. [National Center for Biotechnology Information. PubChem Compound Database; CID=5753].Specifications
| 78.125 to 10,000 pg/mL | |
| Corticosterone Competitive ELISA Kit | |
| Fluorescence | |
| Fecal Extract, Plasma, Serum, Supernatant, Urine | |
| ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti | |
| 7.9% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Fecal Extract 50 μL, Plasma 1 μL, Serum 1 μL, Supernatant 50 μL, Urine 2.5 μL | |
| Corticosterone | |
| 1 hrs 30 mins |
| 18.6 pg/mL | |
| HRP | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| 5.2 | |
| HRP | |
| Inflammation | |
| -20°C | |
| 1 hrs 20 mins |