missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cortexolone 98.0+%, TCI America™
Spécifications
Spécifications
| Quantity | 1 g |
| Formula Weight | 346.47 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cortexolone |
Sécurité et manipulation
| RTECSNumber | TU5011500 |
| TSCA | No |