accessibility menu, dialog, popup

UserName

CAS RN 152-58-9

H3C(R)O(R)(S)(S)H3C(S)(R)HOOHO

CAS RN 152-58-9

IUPAC Name: (1R,3aS,3bR,9aR,9bS,11aS)-1-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: 11-deoxycortisol reichsteins substance S reichsteins substance s substance s, reichstein's 17-α-hydroxycortexone cortexolone reichstein's compound S reichstein substance S reichstein substance s 17,21-hydroxyprogesterone
Molecular Weight (g/mol): 346.47
Molecular Formula: C21H30O4
InChi Key: WHBHBVVOGNECLV-OBQKJFGGSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results