missing translation for 'onlineSavingsMsg'
Learn More

Brady™ CORROSIVE Hazardous Material Shipping Labels

Catalog No. 19110967
Change view
Click to view available options
Dimensions (L x W):
4 x 4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19110967 4 x 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19110967 Supplier Brady™ Supplier No. 121070

Adhesive Vinyl: Permanent acrylic adhesive sticks to a wide variety of surfaces

  • Indoor/outdoor vinyl film withstands exposure to moisture, chemicals and handling.
  • Up to 3 year average outdoor durability

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Description Label
Adhesive Type Self-adhesive
Color Black on White
Format Label
Material Vinyl
Printable Sides Single-side Printable
Product Type Label
Languages English
Quantity 500/Roll
Shape Rectangle
Size (English) 4 x 4 in.
Dimensions (L x W) 4 x 4 in.
Width (English) 4 in.
Width (Metric) 149.86 mm
Height (English) 4 in.
Show More Show Less