missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corosolic acid, ≥85% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 4547-24-4 |
| Physical Form | Neat |
| Linear Formula | C30H48O4 |
| MDL Number | MFCD06794973 |
| Synonym | (2 alpha,3 beta)-2,3-Dihydroxyurs-12-en-28-oic acid; 2 alpha-Hydroxyursolic acid; Colosolic acid; Corsolic acid; Glucosol |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 472.70 |
| Percent Purity | ≥85% (HPLC) |
| Grade | Analytical Standard |