missing translation for 'onlineSavingsMsg'
Learn More

Coronene 83.0+%, TCI America™

Catalog No. C03865G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03865G 5 g
C03861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03865G Supplier TCI America Supplier No. C03865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 191-07-1
Molecular Formula C24H12
Molecular Weight (g/mol) 300.36
MDL Number MFCD00004134
InChI Key VPUGDVKSAQVFFS-UHFFFAOYSA-N
Synonym hexabenzobenzene, dibenzo ghi,pqr perylene, superbenzene, unii-7yy0x5xt1w, ccris 908, 7yy0x5xt1w, 6 circulene, coronene, acmc-209eul
PubChem CID 9115
ChEBI CHEBI:29863
IUPAC Name coronene
SMILES C1=CC2=CC=C3C=CC4=CC=C5C=CC6=CC=C1C1=C2C3=C4C5=C61

Specifications

Melting Point 440°C
Boiling Point 525°C
Quantity 5 g
Formula Weight 300.36
Percent Purity ≥83.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Coronene
RTECSNumber GM5400000
TSCA No