missing translation for 'onlineSavingsMsg'
Learn More

Coronene, 95%

Catalog No. p-93888
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110511000 100 mg Glass bottle
AC110515000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110511000 Supplier Thermo Scientific Chemicals Supplier No. 110511000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 191-07-1 | C24H12 | 300.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 191-07-1
Molecular Formula C24H12
Molecular Weight (g/mol) 300.36
MDL Number MFCD00004134
InChI Key VPUGDVKSAQVFFS-UHFFFAOYSA-N
Synonym hexabenzobenzene, dibenzo ghi,pqr perylene, superbenzene, unii-7yy0x5xt1w, ccris 908, 7yy0x5xt1w, 6 circulene, coronene, acmc-209eul
PubChem CID 9115
ChEBI CHEBI:29863
IUPAC Name coronene
SMILES C1=CC2=CC=C3C=CC4=CC=C5C=CC6=CC=C1C1=C2C3=C4C5=C61

Specifications

Melting Point 438°C to 440°C
Color Green to Yellow
Boiling Point 525°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 mg
Merck Index 15, 2523
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in concentrated sulfuric acid,slightly soluble in benzene
Formula Weight 300.35
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Coronene, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-881-7
RTECSNumber GM5400000

RUO – Research Use Only