Learn More
Corning™ Universal Fit Pipet Tips: Stack Rack, Nonsterile
Designed to assure a reliable fit with major brand pipetters
Supplier: Corning™ C4803
Description
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
| 1 to 200 μL | |
| Natural | |
| Round | |
| 400 Tips x 10 Racks/Cs. | |
| Polypropylene |
| Non-sterile | |
| Rack | |
| 400 Tips/Stack Rack | |
| Yes | |
| 10, 50, 100μL |