missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Universal Fit Pipet Tips: Stack Rack, Nonsterile
GREENER_CHOICE

Catalog No. p-4902643
Change view
Click to view available options
Quantity:
400 Tips x 10 Racks/Cs.
400 Tips x 2 Racks/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07200500 400 Tips x 2 Racks/Cs.
07200581 400 Tips x 10 Racks/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07200500 Supplier Corning™ Supplier No. 4806
Only null left

Designed to assure a reliable fit with major brand pipetters

  • Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
  • Beveled orifice helps ensure accurate fluid delivery.
  • 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
  • Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
  • RNase-/DNase-free.
  • Nonpyrogenic.

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Natural
Format Rack
Tip Style Universal
Packaging 400 Tips/Stack Rack
Quantity 400 Tips x 2 Racks/Cs.
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Graduations 10, 50, 100μL
Show More Show Less