Learn More
Corning™ Universal Fit Pipet Tips: Bulk Packed
Designed to assure a reliable fit with major brand pipetters
Supplier:  Corning™ C4866
 View more versions of this product
Catalog No. 07-200-297
Only null left
Explore available promotions
Description
Description
Designed to assure a reliable fit with major brand pipetters
  • Beveled orifice helps ensure accurate fluid delivery
  • 1–200μL tips are graduated at 10, 50 and 100μL
  • 100–1000μL tips are graduated at 250, 500, and 1000μL
  • Choice of sterile or nonsterile
  • Bulk packed tips are nonsterile and very economical
Specifications
Specifications
1 to 200 μL
Yellow
Round
Universal Fit
Polypropylene
Non-sterile
Bulk
1000 Tips/Bag, 1000 Tips/Cs.
Yes
Product Suggestions
Product Suggestions