missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Ultra-Low Attachment Microplates
GREENER_CHOICE

Catalog No. 07200601
Change view
Click to view available options
Diameter (Metric):
3.48 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
07200601 3.48 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07200601 Supplier Corning™ Supplier No. C3471
Get Up to $2,500 in Products with Purchase
View all promotions
Edge

Featuring a covalently bound hydrogel layer that effectively inhibits cellular attachmment. Corning™ Costar™ Ultra-Low Attachment Microplates greatly reduce binding of attachment and serum proteins, minimizing cell attachment and spreading.

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic

Specifications

Array 2 x 3
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 2.9 mL (Well)
Color Clear
Culture Area 9.5 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Type Multi-well Cell Culture Plate
Well Count 6
For Use With (Application) Maintaining cells in a suspended, unattached state
Includes Lid
No. per Case 24/Cs.
No. per Pack 1/Pk.
Packaging Individually Wrapped
Quantity 24/Cs.
Diameter (Metric) 3.48 cm
Show More Show Less