missing translation for 'onlineSavingsMsg'
Learn More

Corning™ T-Cell Culture Supplement with ConA (IL-2 Culture Supplement), Rat
GREENER_CHOICE

Catalog No. CB40115
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CB40115 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB40115 Supplier Corning™ Supplier No. 354115

Promotes polyclonal activation and IL-2 proliferation of most antigenically sensitized rat or mouse lymphoid T-cells

The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.

  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions

Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.

Order Info

Supplied in 100mL bottles; refrigerate as detailed on label

Specifications

Content And Storage 2° to 8°C
Concentration 10 μg/mL
Product Type T-cell Culture Supplement
Quantity 100 mL
Source Rat