missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Square Polycarbonate Storage Bottles with 45 mm Cap

Catalog No. 361018062
Change view
Click to view available options
Capacity (Metric):
150 mL
250 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
361018062 250 mL
13700400 150 mL
13700412 500 mL
361018064 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 361018062 Supplier Corning™ Supplier No. 431431
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Linerless, one-piece, autoclavable orange GL45 threaded polypropylene plug-seal caps

Space-Saving square shape requires less space (13 to 20%) on the shelf or in the autoclave. Corning™ Square Polycarbonate Storage Bottles are ideal for mixing, sampling and storage.

Corning™ square polycarbonate storage bottles are easier to handle, require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing, sampling and storage.


Caution: These square polycarbonate bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

  • Strong polycarbonate bottles are more break resistant than other glass or plastic bottles.

  • Use a linerless, 45mm one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.

  • Screened white enamel graduations are easier to see than molded graduations

  • Sterilized by gamma radiation

  • Large white marking spot for easier identification

  • Bottles can be autoclaved once at 121oC and 15psi. Repeated autoclaving weakens polycarbonate and is not recommended.

  • Optional reusable caps with silicone septa (Corning Cat. Nos. 1395-45HTSC, 1395-45SS, 1395-45TS) for syringe sampling or introduction of reagents are available for these caps.

  • Note: Most GL45 threaded caps designed for glass storage bottles use a different sealing design and will not give a secure seal if used on these plastic bottles unless they have a rubber liner.
  • Specifications

    Capacity 7.5 oz., 250 mL
    Capacity (Metric) 250 mL
    Material Polycarbonate
    No. per Case 24/Cs.
    No. per Pack 12/Pk.
    Sterility Sterile
    Closure Color Orange
    Graduations 50 to 150 mL
    Closure Material Polypropylene
    Closure Size GL 45
    Closure Type Screw Cap
    Autoclavable Autoclavable
    Capacity (English) 7.5 oz.
    Shape Square
    Show More Show Less