Learn More
Corning™ Reusable Plastic Erlenmeyer Flasks with Red Graduations
Translucent polymethylpentene; with polypropylene screw cap
Supplier: Corning™ 4990P250
Description
Translucent polymethylpentene flasks are manufactured with red graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
Specifications
| Polypropylene | |
| Polypropylene | |
| Narrow Mouth | |
| 1/Pk. |
| 250 mL | |
| 50 mL | |
| 6/Cs. | |
| Screw Cap |