missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PureCoat™ Amine Flasks
Positively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays
$84.92 - $996.52
Specifications
| Closure Material | Polystyrene |
|---|---|
| Material | Polystyrene |
| For Use With (Application) | For preparative cell culture, cell-based and HTS screening assays |
| No. per Pack | 5 Pk. |
| Shape | Rectangular |
| Catalog Number | Mfr. No. | Capacity (Metric) | No. per Case | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | No. per Case | Price | Quantity | ||||
|
08-774-023
|
Corning™
356728 |
750 mL | 40/Cs. |
Case of 40 Each for $996.52
|
|
||||
|
08-774-020
|
Corning™
354726 |
250 mL | 5/Cs. |
Case of 5 Each for $84.92
|
|
||||
|
08-774-021
|
Corning™
356726 |
250 mL | 50/Cs. |
Case of 50 Each for $637.37
|
|
||||
|
08-774-022
|
Corning™
354728 |
750 mL | 5/Cs. |
Case of 5 Each for $180.83
|
|
||||
Description
Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.Specifications
| Polystyrene | |
| For preparative cell culture, cell-based and HTS screening assays | |
| Rectangular | |
| Sterile |
| Polystyrene | |
| 5 Pk. | |
| Amine | |
| Vented |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.