missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PureCoat™ Amine Flasks
Positively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays
Supplier: Corning™ 354726
Description
Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.Specifications
| 8.45 oz. | |
| 250 mL | |
| For preparative cell culture, cell-based and HTS screening assays | |
| 5/Cs. | |
| Rectangular | |
| Sterile | |
| 75 cm2 |
| Polystyrene | |
| Polystyrene | |
| Canted | |
| 5 Pk. | |
| Amine | |
| Vented |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.