Learn More
Corning™ Primaria™ Tissue Culture Dishes
GREENER_CHOICE
Modified polystyrene surface for enhanced cell culture. Corning™ Primaria™ Tissue Culture Dishes feature flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells.
$652.46 - $1,578.10
Specifications
Bottom Shape Flat
Color Clear
Material Polystyrene
Shape Round
Sterility Sterile
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Culture Area Volume (Metric) Working Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Culture Area Volume (Metric) Working Height (Metric) Price Quantity & Availability  
087724A
GREENER_CHOICE
View Documents Promotion Details
Corning™
353801
39.14 mm 9.6 cm2 2.5 to 3 mL 11.05 mm
Case of 200 Each for $652.46
Only null left
087724C
GREENER_CHOICE
View Documents Promotion Details
Corning™
353802
58.8 mm 21.3 cm2 6 to 7 mL 9.53 mm
Case of 200 Each for $728.14
Only null left
087724F
GREENER_CHOICE
View Documents Promotion Details
Corning™
353803
100 mm 58.95 cm2 16 to 17.5 mL 20 mm
Case of 200 Each for $1,578.10
Only null left
Description
Description
The Corning Primaria surface features a unique mixture of oxygen-containing (negatively charged) and nitrogen-containing (positively charged) functional groups on the polystyrene surface. The surface supports the growth of cells that can exhibit poor attachment or limited differentiation potential when cultured on traditional TC surfaces, including neuronal, primary, endothelial, and tumor cells. The surface consistency of each lot is confirmed by electron spectroscopy chemical analysis (ESCA).
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
Order Info
Packaged in red color-coded peel-open, medical-style packages; Available in three sizes; supplied 20 per sleeve in cases of 200; With Lids
Specifications
Specifications
Flat
Polystyrene
Sterile
Cell Culture Dish
20/Pk.
Clear
Round
Multi-treated
200/Cs.